Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) (30 - 32% Ru)

Supplier: Thermo Fisher Scientific
Warning

295782500 295780010
ACRO295782500EA 75.3 EUR
ACRO295782500 ACRO295780010
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) (30 - 32% Ru)
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II)

CAS No.: 12289-94-0

Formula: C₁₆H₂₆Ru
Melting Pt: 81…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216965
CAS Number: 12289-94-0

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 80 to 84 °C
NMR ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR