Hydrazine hydrate 55% in aqueous solution (∼35% w/w N₂H₄)

Supplier: Thermo Fisher Scientific
Danger

296815000 296810025 296811000
ACRO296815000EA 64.9 EUR
ACRO296815000 ACRO296810025 ACRO296811000
Hydrazine hydrate 55% in aqueous solution (∼35% w/w N₂H₄)
Hydrazine hydrate
Formula: NH₂NH₂·xH₂O
Melting Pt: –65 °C
Density: 1,023 g/cm³
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149931
CAS Number: 10217-52-4
EINECS: 206-114-9
UN: 3293
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Solution
Refractive index 1.3860 to 1.3900 (20°C, 589 nm)
Density (20°C) 1.021 to 1.025 g/ml
Titration with HCl 54.5 to 55.5 % (N2H4.xH2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR