4-Fluoro-3-nitrobenzoic acid 98%

Supplier: Thermo Fisher Scientific
Warning

305060050 305060250
ACRO305060050EA 44 EUR
ACRO305060050 ACRO305060250
4-Fluoro-3-nitrobenzoic acid 98%
4-Fluoro-3-nitrobenzoic acid
Formula: C₇H₄FNO₄
Boiling Pt: 92 °C (15 mmHg)
Melting Pt: 120…126 °C
MDL Number: MFCD00007058
CAS Number: 453-71-4
EINECS: 207-221-3

Order Now

Specification Test Results

Appearance Light yellow to tan Powder or crystals
Infrared spectrum Conforms
Melting point 119 to 126 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR