5-Bromo-1-indanone 97%

Supplier: Thermo Fisher Scientific
Warning

309570010 309570050
ACRO309570010EA 44.2 EUR
ACRO309570010 ACRO309570050
5-Bromo-1-indanone 97%
5-Bromo-1-indanone
Formula: C₉H₇BrO
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00082718
CAS Number: 34598-49-7

Order Now

Specification Test Results

Appearance White to beige to brown Powder
Infrared spectrum Conforms
Melting point 125 to 130 °C
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR