Diphenylphosphine oxide 97%

Supplier: Thermo Fisher Scientific
Warning

310120010 310120050
ACRO310120010EA 71 EUR
ACRO310120010 ACRO310120050
Diphenylphosphine oxide 97%
Diphenylphosphine oxide
Formula: C₁₂H₁₁OP
MW: 202.20 g/mol
Melting Pt: 50…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00002079
CAS Number: 4559-70-0

Order Now

Specification Test Results

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
Melting point 50 to 57 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR