4-Methoxyphenyl chloroformate 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Methoxyphenyl carbonochloridate

314570050 314570250
ACRO314570050EA 81.5 EUR
ACRO314570050 ACRO314570250
4-Methoxyphenyl chloroformate 98%
4-Methoxyphenyl chloroformate
Formula: C₇H₄BrClO₂
MW: 186.5924 g/mol
Boiling Pt: 49 °C (.2 mmHg)
Density: 1.25 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013258
CAS Number: 7693-41-6
UN: 3277
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5260 (20°C, 589 nm)
GC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR