Lanthanum(III) chloride 99.9%

Supplier: Thermo Fisher Scientific
Danger

315230100 315230500 315232500
ACRO315230100EA 116 EUR
ACRO315230100 ACRO315230500 ACRO315232500
Lanthanum(III) chloride 99.9%
Lanthanum (III) chloride
Formula: Cl₃La
Boiling Pt: 1000 °C
Melting Pt: 860 °C
Density: 3,8773 g/cm³ (-0.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011068
CAS Number: 10099-58-8
EINECS: 233-237-5
UN: 1759
ADR: 8,III
Merck Index: 13,05379

Order Now

Specification Test Results

Appearance White to off-white Powder
Total conc. of imp.as REO ≤0.1 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR