Iron(II) sulphide 99.9% (trace metals basis) -100 mesh

Supplier: Thermo Fisher Scientific

Synonyms: Ferrous sulphide

315870100 315870500
ACRO315870100EA 101 EUR
ACRO315870100 ACRO315870500
Iron(II) sulphide 99.9% (trace metals basis) -100 mesh
Iron (II) sulphide
Formula: FeS
Melting Pt: 1195 °C
Density: 4,84 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
ADR: 9,III
Merck Index: 12,04106

Order Now

Specification Test Results

Appearance Grey to brown to black Powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR