Tin 99.8%, powder -325 mesh

Supplier: Thermo Fisher Scientific
Warning

317801000 317805000
ACRO317801000EA 54.8 EUR
ACRO317801000 ACRO317805000
Tin 99.8%, powder -325 mesh
Tin
Formula: Sn
Boiling Pt: 2270 °C (760 mmHg)
Melting Pt: 231,9 °C
Density: 7,3 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now

Specification Test Results

Appearance Grey Powder
Total trace metal impurities ≤0.2 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR