Copper 99+%, turnings

Supplier: Thermo Fisher Scientific

317912500 317910010
ACRO317912500EA 70 EUR
ACRO317912500 ACRO317910010
Copper 99+%, turnings
Copper
Formula: Cu
Boiling Pt: 2595 °C (760 mmHg)
Melting Pt: 1083 °C
Density: 8,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Specification Test Results

Appearance Metallic turnings
Total trace metal impurities ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR