3,5-Bis(trifluoromethyl)phenyl isothiocyanate 99+%

Supplier: Thermo Fisher Scientific
Danger

319520010 319520050
ACRO319520010EA 38.3 EUR
ACRO319520010 ACRO319520050
3,5-Bis(trifluoromethyl)phenyl isothiocyanate 99+%
3,5-Bis(trifluoromethyl)phenyl isothiocyanate
Formula: C₉H₃F₆NS
Boiling Pt: 63 °C (1,5 mmHg)
Density: 1,485 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00040838
CAS Number: 23165-29-9
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear to slightly hazy colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5020 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR