Tetrahydropyran 99%

Supplier: Thermo Fisher Scientific
Danger

325881000 325885000
ACRO325881000EA 70.9 EUR
ACRO325881000 ACRO325885000
Tetrahydropyran 99%
Tetrahydropyran
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 88
Melting Pt: –45 °C
Density: 0.88 g/cm³
Flash Pt: –15 °C
MDL Number: MFCD00006585
CAS Number: 142-68-7
EINECS: 205-552-8
UN: 3271
ADR: 3 II
Merck Index: 12,09356

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR