Ammonium peroxodisulphate (APS) 99+% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Fisher Scientific
Danger

327081000 327085000
ACRO327081000EA 48.5 EUR
ACRO327081000 ACRO327085000
Ammonium peroxodisulphate (APS) 99+% for molecular biology DNase-, RNase-, protease-free
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
Melting Pt: 120 °C (dec.)
Density: 1,98 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Titration Iodometric ≥99 %
Acidity ≤0.2 % (H2SO4)
Dnase,Rnase,Protease activity none detected
Chloride (Cl) ≤20 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR