Nω-Methyl-L-arginine monoacetate 99%

Supplier: Thermo Fisher Scientific

Synonyms: H-Arg(Me)-OH.AcOH, N5-(Methylamidino)-L-ornithine acetate salt, Nomega-Monomethyl-L-arginine acetate, L-NMMA, Ng-Methyl-L-Arginine Acetate Salt, Nomega-Methyl-L-arginine acetate, N-ω-Monomethyl-L-arginine monoacetate, NG-Monomethyl-L-arginine acetate salt

328720050 328720250 328721000
ACRO328720050EA 52.9 EUR
ACRO328720050 ACRO328720250 ACRO328721000
Nω-Methyl-L-arginine monoacetate 99%
Nω-Methyl-L-arginine monoacetate

CAS No.: 53308-83-1

Formula: C₇H₁₆N₄O₂·CH₃COOH
Melting Pt: 180…190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00069311
CAS Number: 53308-83-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 180 to 190 °C
TLC ≥98.5 %
Specific optical rotation +9° to +13° (20 °C, 589 nm) (c=4, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR