(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride

Supplier: Thermo Fisher Scientific
Warning

328871000 328875000
ACRO328871000EA 214 EUR
ACRO328871000 ACRO328875000
(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride
(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride
Formula: C₉H₁₅N₅O₃·2HCl
Melting Pt: 250…255 °C
Storage Temperature: Freezer
MDL Number: MFCD00891665
CAS Number: 69056-38-8

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR