Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy

Supplier: Thermo Fisher Scientific
Danger

329920050 329920100 329920500
ACRO329920050EA 118 EUR
ACRO329920050 ACRO329920100 ACRO329920500
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Methanol-[D4]
Formula: D₃COD
Boiling Pt: 65,3 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0,888 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 % (Coulometric)
D-Enrichment ≥99.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR