Diethyl (aminomethyl)phosphonate oxalate 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Phosphonic acid, P-(aminomethyl)-, diethyl ester, ethanedioate (1:1), Diethyl (aminomethyl)phosphonate ethanedioate, diethyl aminomethylphosphonate oxalate

332210010 332210050
ACRO332210010EA 150 EUR
ACRO332210010 ACRO332210050
Diethyl (aminomethyl)phosphonate oxalate 95%
Diethyl (aminomethyl)phosphonate oxalate

CAS No.: 117196-73-3

Formula: C₇H₁₆NO₇P
MW: 257.179 g/mol
Melting Pt: 118…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00800255
CAS Number: 117196-73-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 118 to 126 °C
Titration with HClO4 ≥94 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR