2-Diethoxyphosphorylethylazanium 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Diethyl (2-aminoethyl)phosphonate oxalate

332220010 332220050
ACRO332220010EA 150 EUR
ACRO332220010 ACRO332220050
2-Diethoxyphosphorylethylazanium 97%
2-Diethoxyphosphorylethylazanium

CAS No.: 203192-98-7

Formula: C₆H₁₆NO₃P·C₂H₂O₄
MW: 271.21 g/mol
Melting Pt: 135…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00800256
CAS Number: 203192-98-7

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 135 to 141 °C
Titration with HClO4 ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR