Ammonium iron(III) oxalate trihydrate 98%, pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Iron ammonium oxalate, Ammonium ferric oxalate, Ammonium iron oxalate trihydrate

335100500 335100010 335100051
ACRO335100500EA 42.7 EUR
ACRO335100500 ACRO335100010 ACRO335100051
Ammonium iron(III) oxalate trihydrate 98%, pure
Ammonium iron(III) oxalate trihydrate
Formula: (NH₄)₃Fe(C₂O₄)₃·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00150137
CAS Number: 13268-42-3
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance Green Crystalline powder
Infrared spectrum Conforms
Titration with KMnO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR