4-MUG trihydrate 98%

Supplier: Thermo Fisher Scientific

Synonyms: 4-methylumbelliferyl-beta-D-glucuronide trihydrate

337220010 337220050 337220250
ACRO337220010EA 65 EUR
ACRO337220010 ACRO337220050 ACRO337220250
4-MUG trihydrate 98%
4-MUG trihydrate
Formula: C₁₆H₁₆O₉
MW: 352.29 g/mol
Melting Pt: 102 °C
Storage Temperature: Freezer
MDL Number: MFCD00036772
CAS Number: 6160-80-1

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Infrared spectrum Conforms
Water 8 to 10.2 % (K.F.)
Solubility (c=1, H2O at 60°C) Clear colorless solution
HPLC ≥97.5 %
Specific optical rotation -103° to -107° (20°C, 589 nm) (c=0.25, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR