Dimethyl(2-hydroxyethyl)phosphonate 95%

Supplier: Thermo Fisher Scientific
Warning

338160050 338160250
ACRO338160050EA 128 EUR
ACRO338160050 ACRO338160250
Dimethyl(2-hydroxyethyl)phosphonate 95%
Dimethyl(2-hydroxyethyl)phosphonate

Appearance: Clear colorless to light yellow Liquid

Formula: C₄H₁₁O₄P
Boiling Pt: 98…104 °C (,1 mmHg)
Density: 1,247 g/cm³
Flash Pt: 192 °C
Storage Temperature: Ambient
MDL Number: MFCD00043376
CAS Number: 54731-72-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4460 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR