3-Chlorophenethylamine 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(3-Chlorophenyl)ethylamine

342160050 342160250
ACRO342160050EA 67.2 EUR
ACRO342160050 ACRO342160250
3-Chlorophenethylamine 98%
3-Chlorophenethylamine
Formula: C₈H₁₀ClN
Boiling Pt: 111…113 °C (12 mmHg)
Flash Pt: >109 °C
Storage Temperature: Ambient
MDL Number: MFCD00047957
CAS Number: 13078-79-0
EINECS: 235-982-1

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR