(S)-(+)-Abscisic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: ABA, (+)-cis,trans ABA, (S)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-(2Z,4E)-pentadienoic acid, (S)-(+)-Abscisic acid, Dormin, S-ABA, (+)-Abscisic acid

342401000 342405000
ACRO342401000EA 72.8 EUR
ACRO342401000 ACRO342405000
(S)-(+)-Abscisic acid ≥98%
(S)-(+)-Abscisic acid

  • Bioactive

Formula: C₁₅H₂₀O₄
Melting Pt: 160…163 °C
Storage Temperature: Freezer
MDL Number: MFCD00066545
CAS Number: 21293-29-8

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 158 to 164 °C
Loss on drying ≤2 %
HPLC ≥97.5 %
Specific optical rotation +405° to +413° (20 °C, 589 nm) (c=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR