1-Butanesulphonic acid sodium salt 99+% HPLC

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Sodium 1-butanesulphonate, Sodium butane-1-sulphonate, Sodium 1-butansulphonate, Butane-1-sulphonic acid sodium salt

343800050 343800250
ACRO343800050EA 109 EUR
ACRO343800050 ACRO343800250
1-Butanesulphonic acid sodium salt 99+% HPLC
1-Butansulphonic acid sodium salt
Formula: C₄H₉NaO₃S
MW: 160.17 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1
EINECS: 219-201-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 105°C, 2 hrs)
Solubility (10 % in water) Clear to slightly hazy colorless
UV at 280 nm A: ≤0.04
Titration after Ion exchange ≥99.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR