Disodium hydrogen phosphate dihydrate 99+% for HPLC

Supplier: Thermo Fisher Scientific

343810250 343810025 343811000 343815000
ACRO343810250EA 27.5 EUR
ACRO343810250 ACRO343810025 ACRO343811000 ACRO343815000
Disodium hydrogen phosphate dihydrate 99+% for HPLC
di-Sodium hydrogen phosphate dihydrate
Formula: Na₂HPO₄·2H₂O
Melting Pt: 243…245 °C
Density: 2,1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7
EINECS: 231-448-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
UV at 254 nm A: ≤0.05
pH 7.5 to 9.2 (5 % at 25°C)
Titration with HCl ≥99.0 %
Chloride (Cl) ≤50 ppm
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR