Lanthanum(III) trifluoromethanesulphonate 99%

Supplier: Thermo Fisher Scientific
Danger

343910010 343910050
ACRO343910010EA 37.7 EUR
ACRO343910010 ACRO343910050
Lanthanum(III) trifluoromethanesulphonate 99%
Lanthanum (III) trifluoromethanesulphonate
Formula: C₃F₉LaO₉S₃
Density: 1,7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00077560
CAS Number: 52093-26-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light beige Powder or crystals
Infrared spectrum Conforms
Elemental Analysis 23.35 to 24.05 % (La)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR