Sodium sulphanilate hydrate 97%

Supplier: Thermo Fisher Scientific

346882500
ACRO346882500EA 112 EUR
ACRO346882500
Sodium sulphanilate hydrate 97%
Sodium sulphanilate hydrate

Appearance: White to light brown to pink Crystalline powder

Formula: C₆H₇NO₃S
Melting Pt: 288 °C
Storage Temperature: Ambient
MDL Number: MFCD00064394
CAS Number: 123333-70-0

Order Now

Specification Test Results

Appearance White to light brown to pink Crystalline powder
Infrared spectrum Conforms
Assay ≥96.0 % (on dry substance)
Water ≤16 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR