Allyltrimethylsilane 97%

Supplier: Thermo Fisher Scientific
Danger

349990100 349990500
ACRO349990100EA 54.8 EUR
ACRO349990100 ACRO349990500
Allyltrimethylsilane 97%
Allyltrimethylsilane
Formula: H₂CCHCH₂Si(CH₃)₃
Boiling Pt: 84 °C (1013 hPa)
Density: 0,719 g/cm³
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008635
CAS Number: 762-72-1
EINECS: 212-104-5
UN: 1993

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR