4-Amino-3-hydroxybenzoic acid 98%

Supplier: Thermo Fisher Scientific
Warning

354260050 354260250
ACRO354260050EA 54.8 EUR
ACRO354260050 ACRO354260250
4-Amino-3-hydroxybenzoic acid 98%
4-Amino-3-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00017094
CAS Number: 2374-03-0

Order Now

Specification Test Results

Appearance Yellow-brown to brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR