1-Octyl-3-methylimidazolium hexafluorophosphate 98+%

Supplier: Thermo Fisher Scientific
Warning

356760050 356760250
ACRO356760050EA 87.8 EUR
ACRO356760050 ACRO356760250
1-Octyl-3-methylimidazolium hexafluorophosphate 98+%
1-Methyl-3-octylimidazolium hexafluorophosphate
Formula: C₁₂H₂₃F₆N₂P
MW: 340.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03427617
CAS Number: 304680-36-2

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20°C, 589 nm)
Water ≤1.0 % (K.F.)
NMR (1H-NMR) ≥98.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR