2-Naphthyl boronic acid 97+%

Supplier: Thermo Fisher Scientific
Warning

359620010 359620050
ACRO359620010EA 38.4 EUR
ACRO359620010 ACRO359620050
2-Naphthyl boronic acid 97+%
2-Naphthyl boronic acid
Formula: C₁₀H₉BO₂
Melting Pt: 269…275 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236051
CAS Number: 32316-92-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder and/or chunks
Infrared spectrum Conforms
HPLC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR