Dimethyl sulphoxide-[D₆] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy

Supplier: Thermo Fisher Scientific

360000100 360000500
ACRO360000100EA 78.3 EUR
ACRO360000100 ACRO360000500
Dimethyl sulphoxide-[D₆] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
Boiling Pt: 189 °C (5 mmHg)
Melting Pt: 20,2 °C
Density: 1,19 g/cm³
Flash Pt: 88 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.03 % (Coulometric)
D-Enrichment ≥99.75 %
TMS 0.02 to 0.04 v/v %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR