1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene

Supplier: Thermo Fisher Scientific

Synonyms: 2,4-Dihydro-2,4,5-triphenyl-1,2,4-triazol-3-ylidene, 1,3,4-Triphenyl-4,5-dihydro-1H-triazole-5-ylidene, 2,4,5-Triphenyl-4H-1,2,4-triazol-2-ium-3-ide

363940010
ACRO363940010EA 337 EUR
ACRO363940010
1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene
1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene

Appearance: Beige Crystalline powder

Formula: C₂₀H₁₅N₃
Melting Pt: 150 °C
Storage Temperature: Freezer
MDL Number: MFCD00801868
CAS Number: 166773-08-6

Order Now

Specification Test Results

Appearance Beige Crystalline powder
Infrared spectrum Conforms
NMR Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR