1-Boc-4-hydroxypiperidine 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate

368270050 368270250 368271000
ACRO368270050EA 70.4 EUR
ACRO368270050 ACRO368270250 ACRO368271000
1-Boc-4-hydroxypiperidine 97%
1-Boc-4-hydroxypiperidine
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 60…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01075174
CAS Number: 109384-19-2

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR