Platinum(II) acetylacetonate 98%

Supplier: Thermo Fisher Scientific
Warning

369350010 369350050 369350250
ACRO369350010EA 230 EUR
ACRO369350010 ACRO369350050 ACRO369350250
Platinum(II) acetylacetonate 98%
Platinum (II) acetylacetonate
Formula: C₁₀H₁₄O₄Pt
Melting Pt: 250…252 °C
Density: ~ 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000028
CAS Number: 15170-57-7
EINECS: 239-223-5

Order Now

Specification Test Results

Appearance Yellow to yellow-green Powder
Infrared spectrum Conforms
Assay ≥97.5 % (ex Pt)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR