Strontium chloride, anhydrous 99.99% (trace metals basis)

Supplier: Thermo Fisher Scientific
Danger

369740050 369740250
ACRO369740050EA 124 EUR
ACRO369740050 ACRO369740250
Strontium chloride, anhydrous 99.99% (trace metals basis)
Strontium chloride
Formula: Cl₂Sr
Boiling Pt: 1250 °C (1013 hPa)
Melting Pt: 873 °C
Density: 1,96 g/cm³ (20 °C)
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 12,09000

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay 54.4 to 55.9 % (Sr)
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR