Ethyl benzoylacetate 90%

Supplier: Thermo Fisher Scientific

370850050 370850500 370851000 370855000
ACRO370850050EA 26.5 EUR
ACRO370850050 ACRO370850500 ACRO370851000 ACRO370855000
Ethyl benzoylacetate 90%
Ethyl benzoylacetate
Formula: C₁₁H₁₂O₃
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1,106 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00009196
CAS Number: 94-02-0
EINECS: 202-295-3
Merck Index: 13,03803

Order Now

Specification Test Results

Appearance Clear colorless to yellow to red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5240 to 1.5330 (20°C, 589 nm)
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR