Mexiletine hydrochloride

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-(2,6-Dimethylphenoxy)propan-2-amine hydrochloride, 1-(2,6-Dimethylphenoxy)-2-propylamine Hydrochloride, Mexiletine HCl

372910050 372910250 372911000
ACRO372910050EA 37.2 EUR
ACRO372910050 ACRO372910250 ACRO372911000
Mexiletine hydrochloride
Mexiletine hydrochloride
Formula: C₁₁H₁₇NO
Melting Pt: 202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216024
CAS Number: 5370-01-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 200 to 205 °C
Loss on drying ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR