Dicyclohexyl phthala 99%

Supplier: Thermo Fisher Scientific
Danger

375050050 375052500 375050010 375050025
ACRO375050050EA 36 EUR
ACRO375050050 ACRO375052500 ACRO375050010 ACRO375050025
Dicyclohexyl phthala 99%
Dicyclohexyl phthala
Formula: C₂₀H₂₆O₄
Boiling Pt: 200…235 °C (4 mmHg)
Melting Pt: 63 °C
Flash Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00003849
CAS Number: 84-61-7
EINECS: 201-545-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 63 to 67 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR