Nordihydroguaiaretic acid 95%

Supplier: Thermo Fisher Scientific
Warning

376300010 376300050 376305000
ACRO376300010EA 98 EUR
ACRO376300010 ACRO376300050 ACRO376305000
Nordihydroguaiaretic acid 95%
Nordihydroguaiaretic acid
Formula: C₁₈H₂₂O₄
Melting Pt: 184…189 °C
Storage Temperature: Freezer
MDL Number: MFCD00002206
CAS Number: 500-38-9
EINECS: 207-903-0

Order Now

Specification Test Results

Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Melting point 184 to 189 °C
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR