Cerium(III) carbonate hydrate 99.9% (trace metals basis)

Supplier: Thermo Fisher Scientific
Warning

379400100 379400500 379402500
ACRO379400100EA 26 EUR
ACRO379400100 ACRO379400500 ACRO379402500
Cerium(III) carbonate hydrate 99.9% (trace metals basis)
Cerium (III) carbonate hydrate
Formula: C₃Ce₂O₉·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149635
CAS Number: 54451-25-1
EINECS: 208-655-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Total trace metal impurities ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR