1-Bromo-4-fluoro-3-nitrobenzene 98%

Supplier: Thermo Fisher Scientific
Warning

380890050 380890250
ACRO380890050EA 74.4 EUR
ACRO380890050 ACRO380890250
1-Bromo-4-fluoro-3-nitrobenzene 98%
1-Bromo-4-fluoro-3-nitrobenzene
Formula: C₆H₃BrFNO₂
Boiling Pt: 145…147 °C (35 mmHg)
Melting Pt: 18…20 °C
Density: 1,786 g/cm³
Flash Pt: 110 °C
MDL Number: MFCD00129165
CAS Number: 364-73-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5750 to 1.5780 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR