O-Benzylhydroxylamine 97%

Supplier: Thermo Fisher Scientific
Warning

382050050 382050250 382051000
ACRO382050050EA 53.3 EUR
ACRO382050050 ACRO382050250 ACRO382051000
O-Benzylhydroxylamine 97%
O-Benzylhydroxylamine

CAS No.: 622-33-3

Formula: C₇H₉NO
Boiling Pt: 110…112 °C (20 mmHg)
Density: 1,19 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00221709
CAS Number: 622-33-3

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5410 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR