Samarium(III) trifluoromethanesulphonate 98%

Supplier: Thermo Fisher Scientific
Danger

383170010 383170100
ACRO383170010EA 32 EUR
ACRO383170010 ACRO383170100
Samarium(III) trifluoromethanesulphonate 98%
Samarium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sm
Storage Temperature: Ambient
MDL Number: MFCD00192431
CAS Number: 52093-28-4
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Elemental Analysis 24.65 to 25.66 % (Sm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR