Nitrosium hexafluorophosphate 95%

Supplier: Thermo Fisher Scientific
Danger

383730050 383730250
ACRO383730050EA 133 EUR
ACRO383730050 ACRO383730250
Nitrosium hexafluorophosphate 95%
Nitrosium hexafluorophosphate
Formula: F₆NOP
Storage Temperature: Ambient
MDL Number: MFCD00040324
CAS Number: 16921-91-8
EINECS: 240-980-9
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to white Crystals
Assay ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR