(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine 95%

Supplier: Thermo Fisher Scientific
Warning

386570010 386572500
ACRO386570010EA 210 EUR
ACRO386570010 ACRO386572500
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine 95%
(S)-(-)-3-(tert-Butoxycarbonyl)-4-formyl-2,2-dimethyl-1,3-oxazolidine

CAS No.: 102308-32-7

Formula: C₁₁H₁₉NO₄
Boiling Pt: 83…88 °C (1 mmHg)
Density: 1,06 g/cm³
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209557
CAS Number: 102308-32-7

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4490 (20°C, 589 nm)
GC ≥94 %
Specific optical rotation -85° to -95° (20°C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR