Allyltrimethoxysilane 97+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Trimethoxyallylsilane

388940050 388940250
ACRO388940050EA 59.3 EUR
ACRO388940050 ACRO388940250
Allyltrimethoxysilane 97+%
Allyltrimethoxysilane
Formula: H₂CCHCH₂Si(OCH₃)₃
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00053865
CAS Number: 2551-83-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
GC ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR