Manganese(II) acetate, anhydrous 98+%

Supplier: Thermo Fisher Scientific
Warning

389140050 389140250 389141000
ACRO389140050EA 37.7 EUR
ACRO389140050 ACRO389140250 ACRO389141000
Manganese(II) acetate, anhydrous 98+%
Manganese (II) acetate
Formula: (H₃CCOO)₂Mn
Storage Temperature: Ambient
MDL Number: MFCD00013039
CAS Number: 638-38-0
EINECS: 211-334-3
Merck Index: 12,05763

Order Now

Specification Test Results

Appearance Beige to grey to light brown Powder
Infrared spectrum Conforms
Titration Complexometric ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR