3-Butynyl toluene-4-sulphonate 96%

Supplier: Thermo Fisher Scientific
Warning

390780050 390780250
ACRO390780050EA 112 EUR
ACRO390780050 ACRO390780250
3-Butynyl toluene-4-sulphonate 96%
3-Butynyl toluene-4-sulphonate
Formula: C₁₁H₁₂O₃S
Density: 1,012 g/cm³
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041687
CAS Number: 23418-85-1

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5215 to 1.5255 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR