Triammonium citrate 97+%, pure

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate

391410010 391410500 391412500
ACRO391410010EA 400 EUR
ACRO391410010 ACRO391410500 ACRO391412500
Triammonium citrate 97+%, pure
tri-Ammonium citrate
Formula: C₆H₁₇N₃O₇
Melting Pt: 185 °C
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036406
CAS Number: 3458-72-8

Order Now

Specification Test Results

Appearance White to off-white Crystals
Infrared spectrum Conforms
Titration with HClO4 ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR